N,N-Dimethylformamide dimethyl acetal ≥95%
Supplier: Matrix Scientific
Synonyms:
Esterate M
, N,N-Dimethylformamide Dimethyl Acetal
, DMF-DMA
, 1,1-Dimethoxytrimethylamine
, 1,1-Dimethoxy-N,N-dimethylmethylamine
086025-500G
086025-100G
102846-680EA
63.08
USD
102846-680
102846-682
N,N-Dimethylformamide dimethyl acetal ≥95%
N,N-Dimethylformamide dimethyl acetal
Hazard: Flammable/Moisture Sensitive
Formula:
C₅H₁₃NO₂ MW: 119.16 g/mol Boiling Pt: 102…104 °C Density: 0.897 Flash Pt: 7 °C |
MDL Number:
MFCD00008482 CAS Number: 4637-24-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...