Ethyl-2-chloroacetoacetate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-chloroacetoacetate

086023-100G
102846-678EA 49.23 USD
102846-678
Ethyl-2-chloroacetoacetate ≥97%
Ethyl 2-chloroacetoacetate

Hazard: Flammable/Corrosive

Formula: C₆H₉ClO₃
MW: 164.59 g/mol
Boiling Pt: 107 °C (14 mmHg)
Density: 1.19
MDL Number: MFCD00009141
CAS Number: 609-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR