8-Fluoro-2,3-dihydro-4H-chromen-4-one ≥97%

Supplier: Matrix Scientific

Synonyms: 8-Fluorochroman-4-one

086011-1G
102846-638EA 636.92 USD
102846-638
8-Fluoro-2,3-dihydro-4H-chromen-4-one ≥97%
8-Fluoro-2,3-dihydro-4H-chromen-4-one

Hazard: Irritant

Formula: C₉H₇FO₂
MW: 166.15 g/mol
MDL Number: MFCD09744011
CAS Number: 111141-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR