1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Hydroxy-1,2,4-triazolo[4,3-a]pyridine
, [1,2,4]Triazolo[4,3-a]pyridin-3(2H)-one
086009-50G
086009-10G
102846-634EA
329.23
USD
102846-634
102846-632
1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one ≥97%
1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one
Hazard: Irritant
Formula:
C₆H₅N₃O MW: 135.13 g/mol |
MDL Number:
MFCD00022632 CAS Number: 6969-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...