7-(Trifluoromethyl)-1H-indazole ≥97%

Supplier: Matrix Scientific

086008-1G
102846-630EA 530.77 USD
102846-630
7-(Trifluoromethyl)-1H-indazole ≥97%
7-(Trifluoromethyl)-1H-indazole

Hazard: Irritant

Formula: C₈H₅F₃N₂
MDL Number: MFCD09836171
CAS Number: 885694-00-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR