4-(Trifluoromethyl)-1H-indazole ≥97%

Supplier: Matrix Scientific

086007-1G
102846-628EA 863.08 USD
102846-628
4-(Trifluoromethyl)-1H-indazole ≥97%
4-(Trifluoromethyl)-1H-indazole

Hazard: Irritant

Formula: C₈H₅F₃N₂
MDL Number: MFCD09263211
CAS Number: 1000339-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR