2-Methyl-6-nitroimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

086004-1G
102846-618EA 340 USD
102846-618
2-Methyl-6-nitroimidazo[1,2-a]pyridine ≥97%
2-Methyl-6-nitroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₇N₃O₂
MDL Number: MFCD05863412
CAS Number: 13212-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR