8-Azahypoxanthine ≥97%

Supplier: Matrix Scientific

085996-1G
102846-602EA 863.08 USD
102846-602
8-Azahypoxanthine ≥97%
8-Azahypoxanthine

Hazard: Irritant

Formula: C₄H₃N₅O
MDL Number: MFCD00005804
CAS Number: 2683-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR