2-Methyl-3-nitroimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

085990-1G
102846-586EA 496.92 USD
102846-586
2-Methyl-3-nitroimidazo[1,2-a]pyridine ≥97%
2-Methyl-3-nitroimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₇N₃O₂
MDL Number: MFCD09800612
CAS Number: 34165-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR