3-Nitro-6,7-dihydro-5H-cyclopenta[b]pyridine ≥97%

Supplier: Matrix Scientific

085984-1G
102846-570EA 2333.85 USD
102846-570
3-Nitro-6,7-dihydro-5H-cyclopenta[b]pyridine ≥97%
3-Nitro-6,7-dihydro-5H-cyclopenta[b]pyridine

Hazard: Irritant

Formula: C₈H₈N₂O₂
MDL Number: MFCD09800609
CAS Number: 84531-36-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR