9-Phenylxanthen-9-ol ≥97%

Supplier: Matrix Scientific

Synonyms: 9-Phenyl-9H-xanthen-9-ol

085975-10G
102846-538EA 661.54 USD
102846-538
9-Phenylxanthen-9-ol ≥97%
9-Phenylxanthen-9-ol

Hazard: Irritant

Formula: C₁₉H₁₄O₂
MW: 274.32 g/mol
MDL Number: MFCD00005058
CAS Number: 596-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR