Pyridino[3,2-b]pyrazine ≥97%
Supplier: Matrix Scientific
Synonyms:
Pyrido[2,3-b]pyrazine
085971-1G
102846-526EA
383.08
USD
102846-526
Pyridino[3,2-b]pyrazine ≥97%
Pyridino[3,2-b]pyrazine
Hazard: Irritant
Formula:
C₇H₅N₃ MW: 131.14 g/mol |
MDL Number:
MFCD00006717 CAS Number: 322-46-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...