1-(2'-Fluoro[1,1'-biphenyl]-4-yl)propan-1-ol ≥95%
Supplier: Matrix Scientific
085966-10G
085966-1G
102846-508EA
327.69
USD
102846-508
102846-510
1-(2'-Fluoro[1,1'-biphenyl]-4-yl)propan-1-ol ≥95%
1-(2'-Fluoro[1,1'-biphenyl]-4-yl)propan-1-ol
Hazard: Irritant
Formula:
C₁₅H₁₅FO |
MDL Number:
MFCD00662442 CAS Number: 64820-95-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...