Norborane-2-exo,3-exo-dicarboxylic acid anhydride ≥97%
Supplier: Matrix Scientific
085954-1G
102846-478EA
1118.46
USD
102846-478
Norborane-2-exo,3-exo-dicarboxylic acid anhydride ≥97%
Norborane-2-exo,3-exo-dicarboxylic acid anhydride
Hazard: Irritant
Formula:
C₁₀H₁₂O₃ MW: 180.21 g/mol |
MDL Number:
MFCD11046554 CAS Number: 14166-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...