2,2,5,7-Tetramethylindan-1-one-4-carboxylic acid ≥97%
Supplier: Matrix Scientific
085946-1G
102846-452EA
863.08
USD
102846-452
2,2,5,7-Tetramethylindan-1-one-4-carboxylic acid ≥97%
2,2,5,7-Tetramethylindan-1-one-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₁₆O₃ |
MDL Number:
MFCD00053039 CAS Number: 23342-32-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...