Colchicine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-(-)-Colchicine7α-H-Colchicine

085942-1G
102846-440EA 150.77 USD
102846-440
Colchicine ≥95%
Colchicine

Hazard: Irritant

Formula: C₂₂H₂₅NO₆
MW: 399.44 g/mol
MDL Number: MFCD00078484
CAS Number: 64-86-8
UN: 1544
ADR: 6.1,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR