6-Hydroxy-2-naphthohydrazide ≥97%

Supplier: Matrix Scientific

085932-1G
102846-420EA 806.15 USD
102846-420
6-Hydroxy-2-naphthohydrazide ≥97%
6-Hydroxy-2-naphthohydrazide

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂
MDL Number: MFCD08458497
CAS Number: 849421-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR