6-Hydroxynaphthalene-2-carboximidamide hydrochloride ≥97%
Supplier: Matrix Scientific
085930-1G
102846-414EA
949.23
USD
102846-414
6-Hydroxynaphthalene-2-carboximidamide hydrochloride ≥97%
6-Hydroxynaphthalene-2-carboximidamide hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₁ClN₂O |
MDL Number:
MFCD04039384 CAS Number: 66217-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...