Tricyclazole ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Methyl[1,2,4]triazolo[3,4-b][1,3]benzothiazole
085919-1G
102846-384EA
553.85
USD
102846-384
Tricyclazole ≥97%
Tricyclazole
Hazard: Irritant
Formula:
C₉H₇N₃S |
CAS Number:
41814-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...