Ibudilast ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Isobutyryl-2-isopropylpyrazolo[1,5-a]pyridine1-(2-Isopropylpyrazolo[1,5-a]pyridin-3-yl)-2-methylpropan-1-one

085910-1G
102846-360EA 1118.46 USD
102846-360
Ibudilast ≥97%
Ibudilast

Purity: 97%
CAS: 50847-11-5
MDL Number: MFCD00864808
Molecular Formula: C14H18N2O
Molecular Weight: 230,31
Hazard: Irritant

Formula: C₁₄H₁₈N₂O
MW: 230.31 g/mol
MDL Number: MFCD00864808
CAS Number: 50847-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR