6,7-Dihydro-3H-cyclopenta[d]pyrimidin-4(5H)-one ≥97%

Supplier: Matrix Scientific

085898-1G
102846-326EA 920 USD
102846-326
6,7-Dihydro-3H-cyclopenta[d]pyrimidin-4(5H)-one ≥97%
6,7-Dihydro-3H-cyclopenta[d]pyrimidin-4(5H)-one

Hazard: Irritant

Formula: C₇H₈N₂O
MW: 136.15 g/mol
MDL Number: MFCD00223593
CAS Number: 5661-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR