1-Naphthylacetonitrile ≥97%
Synonyms:
2-(Naphthalen-1-yl)acetonitrile
085883-1G
085883-10G
102846-278EA
301.89
USD
102846-278
102846-276
1-Naphthylacetonitrile ≥97%
1-Naphthylacetonitrile
Hazard: Irritant
Formula:
C₁₂H₉N MW: 167.21 g/mol |
MDL Number:
MFCD00004041 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...