3,3-Dimethyl-2-benzofuran-1(3H)-one ≥97%

Supplier: Matrix Scientific

085882-1G
102846-274EA 750.77 USD
102846-274
3,3-Dimethyl-2-benzofuran-1(3H)-one ≥97%
3,3-Dimethyl-2-benzofuran-1(3H)-one

Hazard: Irritant

Formula: C₁₀H₁₀O₂
MDL Number: MFCD00222552
CAS Number: 1689-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR