4-Methyl-2-benzofuran-1(3H)-one ≥97%
Supplier: Matrix Scientific
085880-1G
102846-270EA
750.77
USD
102846-270
4-Methyl-2-benzofuran-1(3H)-one ≥97%
4-Methyl-2-benzofuran-1(3H)-one
Hazard: Irritant
Formula:
C₉H₈O₂ |
MDL Number:
MFCD11840313 CAS Number: 2211-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...