(1R,5S)-3-Benzyl-3-azabicyclo[3.2.1]octan-8-one ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Benzyl-3-azabicyclo[3.2.1]octan-8-one

085876-1G
102846-260EA 1118.46 USD
102846-260
(1R,5S)-3-Benzyl-3-azabicyclo[3.2.1]octan-8-one ≥97%
(1R,5S)-3-Benzyl-3-azabicyclo[3.2.1]octan-8-one

Hazard: Irritant

Formula: C₁₄H₁₇NO
MW: 215.3 g/mol
MDL Number: MFCD08275096
CAS Number: 83507-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR