2-Methylimidazo[1,2-a]pyridine ≥97%

Supplier: Matrix Scientific

085871-1G
102846-248EA 286.35 USD
102846-248
2-Methylimidazo[1,2-a]pyridine ≥97%
2-Methylimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₈N₂
MW: 132.17 g/mol
MDL Number: MFCD02251330
CAS Number: 934-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR