Bicyclo[4.2.0]octa-1,3,5-triene-7-carbonitrile ≥97%

Supplier: Matrix Scientific

085870-1G
102846-246EA 723.08 USD
102846-246
Bicyclo[4.2.0]octa-1,3,5-triene-7-carbonitrile ≥97%
Bicyclo[4.2.0]octa-1,3,5-triene-7-carbonitrile

Hazard: Irritant

Formula: C₉H₇N
MDL Number: MFCD00001350
CAS Number: 6809-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR