2-Vinylquinoline ≥97%

Supplier: Matrix Scientific

085868-1G
102846-240EA 723.08 USD
102846-240
2-Vinylquinoline ≥97%
2-Vinylquinoline

Hazard: Irritant

Formula: C₁₁H₉N
MW: 155.2 g/mol
CAS Number: 772-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR