tert-Butyl-2-cyclopentylhydrazinecarboxylate ≥97%

Supplier: Matrix Scientific

085849-1G
102846-194EA 467.69 USD
102846-194
tert-Butyl-2-cyclopentylhydrazinecarboxylate ≥97%
tert-Butyl-2-cyclopentylhydrazinecarboxylate

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MW: 200.28 g/mol
MDL Number: MFCD10703555
CAS Number: 646071-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR