1,1'-Biphenyl-4,4'-dicarboxamide ≥97%

Supplier: Matrix Scientific

085832-1G
102846-132EA 863.08 USD
102846-132
1,1'-Biphenyl-4,4'-dicarboxamide ≥97%
1,1'-Biphenyl-4,4'-dicarboxamide

Hazard: Irritant

Formula: C₁₄H₁₂N₂O₂
MDL Number: MFCD00017138
CAS Number: 46902-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR