4,4-Dipropylcyclohex-2-en-1-one ≥97%

Supplier: Matrix Scientific

085825-1G
102846-112EA 226.15 USD
102846-112
4,4-Dipropylcyclohex-2-en-1-one ≥97%
4,4-Dipropylcyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₂H₂₀O
MDL Number: MFCD00269983
CAS Number: 60729-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR