3-Phenoxythiophene ≥97%
Supplier: Matrix Scientific
085813-1G
085813-10G
102846-064EA
241.54
USD
102846-064
102846-066
3-Phenoxythiophene ≥97%
3-Phenoxythiophene
Hazard: Irritant
Formula:
C₁₀H₈OS MW: 176.23 g/mol |
MDL Number:
MFCD00178747 CAS Number: 63285-84-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...