2-Methyl-1,3-dithiane ≥98%
Supplier: Matrix Scientific
085811-1G
085811-10G
085811-50G
102846-052EA
101.54
USD
102846-052
102846-054
102846-056
2-Methyl-1,3-dithiane ≥98%
2-Methyl-1,3-dithiane
Hazard: Irritant
Formula:
C₅H₁₀S₂ |
MDL Number:
MFCD00014652 CAS Number: 6007-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...