1,3-Dithiolane ≥97%

Supplier: Matrix Scientific

085810-10G
102846-048EA 807.69 USD
102846-048
1,3-Dithiolane ≥97%
1,3-Dithiolane

Hazard: Irritant

Formula: C₃H₆S₂
Boiling Pt: 175 °C
Melting Pt: –50 °C
Density: 1.259/17 °C
MDL Number: MFCD00041425
CAS Number: 4829-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR