3,4-Dianilinocyclobut-3-ene-1,2-dione ≥97%

Supplier: Matrix Scientific

085809-1G
102846-044EA 324.62 USD
102846-044
3,4-Dianilinocyclobut-3-ene-1,2-dione ≥97%
3,4-Dianilinocyclobut-3-ene-1,2-dione

Hazard: Irritant

Formula: C₁₆H₁₂N₂O₂
Melting Pt: 265…270 °C
MDL Number: MFCD00179353
CAS Number: 33512-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR