2-Heptylcyclopentan-1-one oxime ≥97%

Supplier: Matrix Scientific

085808-10G 085808-1G
102846-040EA 241.54 USD
102846-040 102846-042
2-Heptylcyclopentan-1-one oxime ≥97%
2-Heptylcyclopentan-1-one oxime

Hazard: Irritant

Formula: C₁₂H₂₃NO
MDL Number: MFCD09998437
CAS Number: 165385-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR