3-Cyclopentene-1-ol ≥95%

Supplier: Matrix Scientific

Synonyms: Cyclopent-3-enol

085803-1G
102846-024EA 106.15 USD
102846-024
3-Cyclopentene-1-ol ≥95%
3-Cyclopentene-1-ol

Hazard: Irritant

Formula: C₅H₈O
MW: 84.12 g/mol
MDL Number: MFCD00798186
CAS Number: 14320-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR