2,6-Dimethyltetrahydro-2H-pyran-4-ol ≥97%

Supplier: Matrix Scientific

085780-1G
102845-970EA 806.15 USD
102845-970
2,6-Dimethyltetrahydro-2H-pyran-4-ol ≥97%
2,6-Dimethyltetrahydro-2H-pyran-4-ol

Hazard: Irritant

Formula: C₇H₁₄O₂
MDL Number: MFCD18089272
CAS Number: 33747-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR