Tetrahydrothiopyran-4-carbonitrile ≥97%

Supplier: Matrix Scientific

085777-1G
102845-960EA 467.69 USD
102845-960
Tetrahydrothiopyran-4-carbonitrile ≥97%
Tetrahydrothiopyran-4-carbonitrile

Hazard: Irritant

Formula: C₆H₉NS
MW: 127.21 g/mol
MDL Number: MFCD09025909
CAS Number: 195503-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR