2-Ethyl-1,3-dioxolane ≥97%

Supplier: Matrix Scientific

085776-1G
102845-958EA 806.15 USD
102845-958
2-Ethyl-1,3-dioxolane ≥97%
2-Ethyl-1,3-dioxolane

Hazard: Irritant

Formula: C₅H₁₀O₂
MDL Number: MFCD00792514
CAS Number: 2568-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR