1-Oxiran-2-ylethanone ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Acetyloxirane

085759-1G
102845-920EA 256.92 USD
102845-920
1-Oxiran-2-ylethanone ≥97%
1-Oxiran-2-ylethanone

Hazard: Irritant

Formula: C₄H₆O₂
MW: 86.09 g/mol
MDL Number: MFCD00829205
CAS Number: 4401-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR