1,2,4,5-Tetrazinane-3,6-dithione ≥97%

Supplier: Matrix Scientific

085755-1G
102845-908EA 750.77 USD
102845-908
1,2,4,5-Tetrazinane-3,6-dithione ≥97%
1,2,4,5-Tetrazinane-3,6-dithione

Hazard: Irritant

Formula: C₂H₄N₄S₂
MDL Number: MFCD00175998
CAS Number: 36239-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR