4-Isopropyl-5-mercapto-4H-1,2,4-triazol-3-ol ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Hydroxy-4-isopropyl-3-sulphanyl-4H-1,2,4-triazole

085738-1G
102845-860EA 553.85 USD
102845-860
4-Isopropyl-5-mercapto-4H-1,2,4-triazol-3-ol ≥97%
4-Isopropyl-5-mercapto-4H-1,2,4-triazol-3-ol

Hazard: Irritant

Formula: C₅H₉N₃OS
MW: 159.21 g/mol
MDL Number: MFCD00068125
CAS Number: 53065-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR