5-Mercapto-4-methyl-4H-1,2,4-triazol-3-ol ≥97%

Supplier: Matrix Scientific

085737-1G
102845-858EA 553.85 USD
102845-858
5-Mercapto-4-methyl-4H-1,2,4-triazol-3-ol ≥97%
5-Mercapto-4-methyl-4H-1,2,4-triazol-3-ol

Hazard: Irritant

Formula: C₃H₅N₃OS
MDL Number: MFCD00068126
CAS Number: 22244-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR