4-Benzyl-1,2,5-thiadiazol-3-ol ≥97%

Supplier: Matrix Scientific

085732-1G
102845-848EA 863.08 USD
102845-848
4-Benzyl-1,2,5-thiadiazol-3-ol ≥97%
4-Benzyl-1,2,5-thiadiazol-3-ol

Hazard: Irritant

Formula: C₉H₈N₂OS
MDL Number: MFCD08235270
CAS Number: 5933-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR