1-Ethyl-1H-1,2,4-triazole ≥97%
Supplier: Matrix Scientific
Synonyms:
1-Ethyl-1,2,4-triazole
085730-1G
102845-844EA
636.92
USD
102845-844
1-Ethyl-1H-1,2,4-triazole ≥97%
1-Ethyl-1H-1,2,4-triazole
Hazard: Irritant
Formula:
C₄H₇N₃ MW: 97.12 g/mol |
MDL Number:
MFCD07367971 CAS Number: 16778-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...