2,2,2-Trichloro-1-(1-methyl-1H-1,2,4-triazol-5-yl)ethanone ≥97%

Supplier: Matrix Scientific

085729-1G
102845-842EA 467.69 USD
102845-842
2,2,2-Trichloro-1-(1-methyl-1H-1,2,4-triazol-5-yl)ethanone ≥97%
2,2,2-Trichloro-1-(1-methyl-1H-1,2,4-triazol-5-yl)ethanone

Hazard: Irritant

Formula: C₅H₄Cl₃N₃O
MDL Number: MFCD11100145
CAS Number: 131758-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR