2-(1H-Pyrrol-1-yl)-1,3,4-thiadiazole ≥97%

Supplier: Matrix Scientific

085728-1G
102845-840EA 723.08 USD
102845-840
2-(1H-Pyrrol-1-yl)-1,3,4-thiadiazole ≥97%
2-(1H-Pyrrol-1-yl)-1,3,4-thiadiazole

Hazard: Irritant

Formula: C₆H₅N₃S
MDL Number: MFCD06653492
CAS Number: 881041-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR