5-(3-Methylbutylthio)-1,3,4-thiadiazol-2-thiol ≥97%

Supplier: Matrix Scientific

085727-1G
102845-838EA 949.23 USD
102845-838
5-(3-Methylbutylthio)-1,3,4-thiadiazol-2-thiol ≥97%
5-(3-Methylbutylthio)-1,3,4-thiadiazol-2-thiol

Hazard: Irritant

Formula: C₇H₁₂N₂S₃
MDL Number: MFCD03789544
CAS Number: 344926-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR