((2-Morpholin-4-ylethyl)amino)(phenylamino)methane-1-thione ≥95%

Supplier: Matrix Scientific

085710-1G
102845-784EA 666.15 USD
102845-784
((2-Morpholin-4-ylethyl)amino)(phenylamino)methane-1-thione ≥95%
((2-Morpholin-4-ylethyl)amino)(phenylamino)methane-1-thione

Hazard: Irritant

Formula: C₁₃H₁₉N₃OS
MDL Number: MFCD01185502
CAS Number: 57723-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR